Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 8.0

Catalog No. AAJ60825AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60825AK 250 mL
AAJ60825AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60825AK Supplier Thermo Scientific Chemicals Supplier No. J60825AK
  • $173.10
    $134.65 / Each of 1

    Save $38.45

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

Solubility
Soluble in water (121 mg/ml at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong acids.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium phosphate buffer soln.
Concentration 0.5 M
pH 8.0
Physical Form Liquid
Quantity 250 mL
MDL Number MFCD00146206
Solubility Information Soluble in water (121mg/ml at 20°C).

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.