Learn More
Electron Microscopy Sciences Sodium Phosphate 1% Aqueous 500 ML
SDP
Catalog No. 5031844
Encompass_Preferred
Used in Fat Stains in Supersaturated Alcohol for fat and lipid stain. See Lillie, R.D.,Histopathologic Technical and Practical Histochemistry, 3rd edition, McGraw-Hill, NY, 1965 or Clark, G., (ed) Staining Procedures, 3rd edition, Williams & Wilkins, Baltimore, p. 149, c. 1973
Catalog No. 50-318-44 Supplier Electron Microscopy Sciences Supplier No. 2650306
Add to Cart
Add to Cart
Used in Fat Stains in Supersaturated Alcohol for fat and lipid stain. See Lillie, R.D.,Histopathologic Technical and Practical Histochemistry, 3rd edition, McGraw-Hill, NY, 1965 or Clark, G., (ed) Staining Procedures, 3rd edition, Williams & Wilkins, Baltimore, p. 149, c. 1973