Learn More
Boston Bioproducts Inc Sodium Phosphate Buffer (0.5 M, pH 7.0) - 500ml
Catalog No. NC0697776
Encompass
The Sodium Phosphate Buffer is a versatile reagent with a high buffering capacity, commonly used across various scientific disciplines across biochemistry and molecular biology. This buffer has a pH range of 5.8 to 8.0. It's applications include protein analysis, where it supports solubility, stability, and activity. It is also compatible with biologically relevant metal ions that are significant for cellular or physiological conditions.
Catalog No. NC0697776 Supplier Boston Bioproducts Inc Supplier No. BB2318500ML
The Sodium Phosphate Buffer is a versatile reagent with a high buffering capacity, commonly used across various scientific disciplines across biochemistry and molecular biology. This buffer has a pH range of 5.8 to 8.0. It's applications include protein analysis, where it supports solubility, stability, and activity. It is also compatible with biologically relevant metal ions that are significant for cellular or physiological conditions.