Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Sodium Phosphate Buffer

Catalog No. MP116560605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116560605 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116560605 Supplier MP Biomedicals™ Supplier No. 116560605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Phosphate Buffer is a reagent with very high buffering capacity that is widely used in molecular biology, biochemistry, and chromatography. It occurs in several forms: monobasic (NaH2PO4), dibasic (Na2HPO4), and tribasic (Na3PO4).

  • Component of the FastDNA SPIN Kit for Soil and FastDNA SPIN Kit for Feces.
  • Sodium phosphate is a reagent with very high buffering capacity that is widely used in molecular biology, biochemistry, and chromatography.
  • It occurs in several forms: monobasic (NaH2PO4), dibasic (Na2HPO4), and tribasic (Na3PO4).
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature (15°C to 30°C)
For Use With (Application) Sample Preparation, Sample Lysis, Nucleic Acid Isolation and Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation
For Use With (Equipment) FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1
pH 7.8 to 8.2
Product Type Reagents
Quantity 500 mL

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.