Learn More
Boston Bioproducts Inc Sodium Phosphate Buffer (20 mM, pH 6.5, HPCE Grade) - 5x50ml
Supplier:  Boston Bioproducts Inc BBP65HPCE5X50ML
Encompass
The HPCE-Grade Sodium Phosphate Buffer is a versatile reagent with a high buffering capacity, commonly used across various scientific disciplines across biochemistry and molecular biology. This buffer has a pH range of 5.8 to 8.0. It's applications include protein analysis, where it supports solubility, stability, and activity. It is also compatible with biologically relevant metal ions that are significant for cellular or physiological conditions.
Catalog No. NC2827787