Learn More
Medchemexpress LLC Sodium phosphate dib 10g
SDP
Catalog No. 5000205837
Encompass_Preferred
Sodium phosphate dibasic dihydrate for molecular biology is an inorganic salt compound Sodium phosphate dibasic dihydrate for molecular biology is commonly used in molecular biology experiments and can be used as a buffer nutritional supplement and detergent
Catalog No. 50-002-05837 Supplier Medchemexpress LLC Supplier No. HYW094708C10G
Sodium phosphate dibasic dihydrate for molecular biology is an inorganic salt compound Sodium phosphate dibasic dihydrate for molecular biology is commonly used in molecular biology experiments and can be used as a buffer nutritional supplement and detergent