Learn More
Medchemexpress LLC Sodium phosphate dib 25g
SDP
Catalog No. 5000206372
Encompass_Preferred
Sodium phosphate dibasic dihydrate for molecular biology is an inorganic salt compound Sodium phosphate dibasic dihydrate for molecular biology is commonly used in molecular biology experiments and can be used as a buffer nutritional supplement and detergent
Catalog No. 50-002-06372 Supplier Medchemexpress LLC Supplier No. 25GHYW094708C
Sodium phosphate dibasic dihydrate for molecular biology is an inorganic salt compound Sodium phosphate dibasic dihydrate for molecular biology is commonly used in molecular biology experiments and can be used as a buffer nutritional supplement and detergent