Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium phosphate dibasic, 7558-79-4, MFCD00003496, 500 g
Catalog No. NC0694078
Encompass
Linear Formula: Na2HPO4, Molecular Weight: 141.96, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, Synonym: sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate.
Catalog No. NC0694078 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S5136500G
Linear Formula: Na2HPO4, Molecular Weight: 141.96, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, Synonym: sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate.