Learn More
Research Products International Corp SODIUM PHOSPHATE DIBASIC HEPTAHYDRATE, ACS GRADE, 5 KILOGRAM
SDP
Supplier:  Research Products International Corp S231755000.0
Encompass_Preferred
Sodium Phosphate Dibasic Heptahydrate, also known as disodium hydrogen phosphate heptahydrate, is a chemical compound with significant applications in the life science marketplace. It is a hydrated form of sodium phosphate dibasic, meaning it contains seven water molecules per molecule of the compound.
Catalog No. 50-997-972