Learn More
Medchemexpress LLC Sodium phosphate dibasic | 7558-79-4 | 99.2% | 141.96 | Na2HPO4 | 10g
SDP
Catalog No. 5000269991
Encompass_Preferred
Sodium phosphate dibasic (Disodium hydrogen phosphate) for cell culture is a buffer and chelating agent Sodium phosphate dibasic is used to adjust the pH of liquids Sodium phosphate dibasic can be used in cell culture[1]
Catalog No. 50-002-69991 Supplier Medchemexpress LLC Supplier No. HYY0308E10G
Sodium phosphate dibasic (Disodium hydrogen phosphate) for cell culture is a buffer and chelating agent Sodium phosphate dibasic is used to adjust the pH of liquids Sodium phosphate dibasic can be used in cell culture[1]