Learn More
Alkali Scientific Sodium Phosphate Dibasic, Anhydrous, 25 Kg
SDP
Catalog No. 5000659943
Encompass_Preferred
Sodium Phosphate Dibasic, Anhydrous, 25 Kg is a high-quality reagent used for various laboratory applications. Sodium Phosphate Dibasic is commonly used in buffer solutions to maintain a stable pH environment in chemical and biological experiments. The anhydrous form is preferred when precise concentrations are necessary for applications such as chromatography, molecular biology, and industrial processes. This large 25 Kg quantity makes it suitable for high-throughput laboratories or production settings. Sodium phosphate dibasic serves as an essential buffer component, ensuring proper pH conditions for enzyme reactions, protein purification, and other biochemical applications. It is widely used in the preparation of standard laboratory reagents, cleaning solutions, and in pharmaceutical testing protocols.
Catalog No. 50-006-59943 Supplier Alkali Scientific Supplier No. EDI002CP
Sodium Phosphate Dibasic, Anhydrous, 25 Kg is a high-quality reagent used for various laboratory applications. Sodium Phosphate Dibasic is commonly used in buffer solutions to maintain a stable pH environment in chemical and biological experiments. The anhydrous form is preferred when precise concentrations are necessary for applications such as chromatography, molecular biology, and industrial processes. This large 25 Kg quantity makes it suitable for high-throughput laboratories or production settings. Sodium phosphate dibasic serves as an essential buffer component, ensuring proper pH conditions for enzyme reactions, protein purification, and other biochemical applications. It is widely used in the preparation of standard laboratory reagents, cleaning solutions, and in pharmaceutical testing protocols.