Learn More
Alkali Scientific Sodium Phosphate Dibasic, Anhydrous, 3 Kg
SDP
Catalog No. 5000659944
Encompass_Preferred
Sodium Phosphate Dibasic, Anhydrous, 3 Kg is a versatile laboratory reagent commonly used for buffering solutions and maintaining pH levels in biological and chemical assays. The anhydrous form of sodium phosphate dibasic ensures that there is no water content, which allows for more accurate and controlled concentrations. This product is used in various applications including chromatography, enzyme assays, and molecular biology techniques where maintaining a stable pH is essential. It is also employed in pharmaceutical and food testing, as well as in preparation of standard solutions for laboratory use. The 3 Kg size is ideal for medium-scale laboratories or for specialized, smaller-scale projects.
Catalog No. 50-006-59944 Supplier Alkali Scientific Supplier No. EDI003CP
Sodium Phosphate Dibasic, Anhydrous, 3 Kg is a versatile laboratory reagent commonly used for buffering solutions and maintaining pH levels in biological and chemical assays. The anhydrous form of sodium phosphate dibasic ensures that there is no water content, which allows for more accurate and controlled concentrations. This product is used in various applications including chromatography, enzyme assays, and molecular biology techniques where maintaining a stable pH is essential. It is also employed in pharmaceutical and food testing, as well as in preparation of standard solutions for laboratory use. The 3 Kg size is ideal for medium-scale laboratories or for specialized, smaller-scale projects.