Learn More
Alkali Scientific Sodium Phosphate Dibasic, Anhydrous, 500 G
SDP
Catalog No. 5000659849
Encompass_Preferred
Sodium Phosphate Dibasic, Anhydrous, 500 G is a reagent used primarily in laboratory applications for preparing buffer solutions and adjusting pH levels. The anhydrous form allows for greater accuracy in concentration control, making it ideal for high-precision experiments and applications in fields such as molecular biology, biochemistry, and analytical chemistry. This medium-size 500 G container is perfect for smaller laboratories, specialized research, and educational purposes. Sodium phosphate dibasic plays a key role in ensuring consistent pH conditions during chemical reactions, enzyme assays, and pharmaceutical testing. It is also used in protein purification, HPLC, and other applications requiring stable buffering.
Catalog No. 50-006-59849 Supplier Alkali Scientific Supplier No. EDI004CP
Sodium Phosphate Dibasic, Anhydrous, 500 G is a reagent used primarily in laboratory applications for preparing buffer solutions and adjusting pH levels. The anhydrous form allows for greater accuracy in concentration control, making it ideal for high-precision experiments and applications in fields such as molecular biology, biochemistry, and analytical chemistry. This medium-size 500 G container is perfect for smaller laboratories, specialized research, and educational purposes. Sodium phosphate dibasic plays a key role in ensuring consistent pH conditions during chemical reactions, enzyme assays, and pharmaceutical testing. It is also used in protein purification, HPLC, and other applications requiring stable buffering.