Learn More
Alkali Scientific Sodium Phosphate Dibasic, Anhydrous, ACS Grade, 5 Kg
SDP
Catalog No. 5000659799
Encompass_Preferred
Sodium Phosphate Dibasic, Anhydrous, ACS Grade, 5 Kg is a high-grade reagent used to prepare phosphate buffers for a variety of laboratory applications. The anhydrous form ensures it remains stable and is easily dissolved to create solutions with precise concentrations. It is commonly used in analytical chemistry and biochemistry for processes requiring a stable pH environment, such as in chromatography, electrophoresis, and cell culture preparation. With a larger 5 Kg packaging, this product is ideal for high-volume laboratories and research facilities, providing an efficient and cost-effective solution for maintaining buffer systems during scientific experimentation.
Catalog No. 50-006-59799 Supplier Alkali Scientific Supplier No. EDI009CP
Sodium Phosphate Dibasic, Anhydrous, ACS Grade, 5 Kg is a high-grade reagent used to prepare phosphate buffers for a variety of laboratory applications. The anhydrous form ensures it remains stable and is easily dissolved to create solutions with precise concentrations. It is commonly used in analytical chemistry and biochemistry for processes requiring a stable pH environment, such as in chromatography, electrophoresis, and cell culture preparation. With a larger 5 Kg packaging, this product is ideal for high-volume laboratories and research facilities, providing an efficient and cost-effective solution for maintaining buffer systems during scientific experimentation.