Learn More
Sodium Phosphate Dibasic Anhydrous (Powder/Laboratory), Fisher Chemical
Shop All Fisherbrand Products

Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Color | White |
| pH | 8.7 to 9.3 |
| Physical Form | Powder/Solid |
| Quantity | 3 kg |
| Identification | Pass Test |
| Loss on Drying | 5% max. |
| Packaging | Poly Bottle |
| Decomposition Information | Decomposes information: > 240°C |
| Formula Weight | 141.96 |
| Grade | Laboratory |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: CAUTION! |
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Hygroscopic. NFPA Health:1 Flammability:0 Instability:1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.