Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phosphate dibasic dodecahydrate, meets analytical spec. of BP, Ph. Eur., E339, 98.5-101% (calc. to the dried substance), Solstice

Catalog No. 60044550 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-550 6 x 2.5 kg
60-018-39 1 kg
60-018-40 2.5 kg
60-044-549 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-044-550 Supplier Solstice Supplier No. 042736X2.5KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

meets analytical specification of BP, European Pharmacopoeia, E339, 98.5-101% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10039-32-4
Molecular Formula H25Na2O16P
Molecular Weight (g/mol) 358.137
MDL Number MFCD00149181
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
Synonym disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
PubChem CID 61456
IUPAC Name disodium;hydrogen phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]

Specifications

Physical Form Crystals
Quantity 6 x 2.5 kg
Linear Formula Na2HPO4 · 12H2O
Formula Weight 358.14
Percent Purity 98.5 to 101% (calc. to the dried substance)
Grade Reagent
Chemical Name or Material Sodium Phosphate Dibasic Dodecahydrate, Meets analytical specification of Ph.Eur.BP
EINECSNumber 231-448-7
RTECSNumber TC5725000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.