Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium phosphate dibasic heptahydrate | 7782-85-6 | MFCD00149180 | 250 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S2429250G
Encompass
Sodium phosphate dibasic heptahydrate | Purity: 99% | Mol Wt: 268.07 | 7782-85-6 | MFCD00149180 | 250 g
Catalog No. NC1467066