Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Phosphate Dibasic Heptahydrate

Catalog No. 43147850G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-147-850G 50 g
43-147-8250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-147-850G Supplier Sigma Aldrich Supplier No. 43147850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium phosphate dibasic heptahydrate has been used to prepare sodium phosphate buffer and phosphate-buffered saline (PBS) buffer.[44}

Specifications

CAS 7782-85-6
Quantity 50 g
Molecular Formula HNa2O4P -+ 7H2O
Linear Formula Na2HPO4 -+ 7H2O
MDL Number MFCD00149180
Synonym Disodium hydrogen phosphate heptahydrate; Disodium phosphate
Molecular Weight (g/mol) 268.07 g/mol
Density 1.68 g/mL at 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.