Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Dibasic Heptahydrate, ACS Reagent (Disodium Hydrogen Phosphate Heptahydrate), Reagents

Catalog No. 01337697
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-697 500 g
01-337-699 10 kg
01-337-698 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-697 Supplier Reagents Holdings Llc Supplier No. C1324000500C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Phosphate Dibasic Heptahydrate, ACS Reagent (Disodium Hydrogen Phosphate Heptahydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate, ACS Reagent (Disodium Hydrogen Phosphate Heptahydrate)
CAS 7782-85-6
Color Colorless/White
Physical Form Solid
Quantity 500 g
Molecular Formula Na2HPO4, 7H2O
Synonym Disodium Hydrogen Phosphate Heptahydrate
Molecular Weight (g/mol) 268.0658
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.