Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Dibasic, MP Biomedicals™

Catalog No. MP021914405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021914405 500 g
MP021914401 100 g
MP219144005 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP021914405 Supplier MP Biomedicals, Inc Supplier No. 02191440.5
Add to Cart
Add to Cart

Widely used in molecular biology, analytical chemistry, biochemistry and chromatography. Sodium Phosphate Dibasic, MP Biomedicals is a reagent with very high buffering capacity, often used in conjunction with Sodium Phosphate, Monobasic in the preparation

Sodium Phosphate Dibasic is used in many applications, including the purification of antibodies, as a reagent and buffer in analytical chemistry, and in the preparation of biological buffers. Also used as a sequestrant, emulsifier and buffer in foods; as mordant in dyeing; for weighting silk; in tanning; in manufacturing of enamels, ceramics, detergents, and boiler compounds; as a fireproofing agent in soldering and brazing instead of borax; as a cathartic; and as a laxative.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Solubility Information Soluble in water, but more soluble in hot water; insoluble in alcohol, ethanol.
IUPAC Name disodium hydrogen phosphate
Formula Weight 141.96
Percent Purity ≥99.0%
Grade ACS
Recommended Storage Room Temperature (15-30°C), dessicate.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.