Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium phosphate monobasic, 7558-80-7, MFCD00003527, 500g
Catalog No. NC1589140
Encompass
Linear Formula NaH2PO4, ≥99.0%, Molecular Weight 119.98, BioXtra, Synonym: Monosodium dihydrogen orthophosphate, Monosodium phosphate, Sodium dihydrogen phosphate.
Catalog No. NC1589140 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S8282500G
Linear Formula NaH2PO4, ≥99.0%, Molecular Weight 119.98, BioXtra, Synonym: Monosodium dihydrogen orthophosphate, Monosodium phosphate, Sodium dihydrogen phosphate.