Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Phosphate Monobasic
SCI_ED

Catalog No. S25562A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Reagent Grade - Crystals
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25562A 100 g Reagent Grade - Crystals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25562A

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
IUPAC Name sodium phosphoric acid hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

Quantity 100 g
Assay Percent Range >98%
Linear Formula NaH2PO4·H2O
Formula Weight 137.99
Grade Reagent
Packaging Reagent Grade - Crystals
Density 1103g/L (20°C)
Chemical Name or Material Sodium Phosphate, Monobasic
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.