Learn More
Alkali Scientific Sodium Phosphate, Monobasic, Anhydrous, 10 Kg
SDP
Catalog No. 5000660992
Encompass_Preferred
Sodium Phosphate Monobasic Anhydrous (10 Kg) is a high-purity reagent used in laboratory applications. As a monobasic phosphate, this anhydrous form is ideal for preparing phosphate buffer solutions, controlling pH in a variety of biochemical processes, and for use in chemical synthesis. This large 10 Kg packaging is suited for high-volume labs requiring consistent phosphate buffering capacity, especially in processes that require stable pH conditions. It is commonly used in the preparation of buffers for chromatography, molecular biology, and analytical chemistry. Sodium phosphate monobasic anhydrous is also used in DNA extraction procedures, enzyme assays, and various other laboratory applications where precise pH regulation is essential.
Catalog No. 50-006-60992 Supplier Alkali Scientific Supplier No. SOA002CP
Sodium Phosphate Monobasic Anhydrous (10 Kg) is a high-purity reagent used in laboratory applications. As a monobasic phosphate, this anhydrous form is ideal for preparing phosphate buffer solutions, controlling pH in a variety of biochemical processes, and for use in chemical synthesis. This large 10 Kg packaging is suited for high-volume labs requiring consistent phosphate buffering capacity, especially in processes that require stable pH conditions. It is commonly used in the preparation of buffers for chromatography, molecular biology, and analytical chemistry. Sodium phosphate monobasic anhydrous is also used in DNA extraction procedures, enzyme assays, and various other laboratory applications where precise pH regulation is essential.