Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Monobasic, Anhydrous, Purified (Sodium Dihydrogen Phosphate), Reagents

Catalog No. 01336988
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-988 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-988 Supplier Reagents Holdings Llc Supplier No. C2323500500A5
Add to Cart
Add to Cart

Sodium Phosphate Monobasic, Anhydrous, Purified (Sodium Dihydrogen Phosphate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
Synonym Sodium Dihydrogen Phosphate
IUPAC Name sodium dihydrogen phosphate
SMILES [Na+].OP(O)([O-])=O

Specifications

Boiling Point 204°C
Color White
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.04
Chemical Name or Material Sodium Phosphate Monobasic, Anhydrous, Purified (Sodium Dihydrogen Phosphate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.