Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium phosphate monobasic BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, >=99.0% (titration) | 7558-80-7 | MFCD00003527 | 500G
Catalog No. 501848807
Encompass
Sodium phosphate monobasic BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, >=99.0% (titration) | Purity: >=99.0% (titration) | Mol Wt: 119.98 | 7558-80-7 | MFCD00003527 | 500G
Catalog No. 501848807 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S5011500G
Add to Cart
Add to Cart
Sodium phosphate monobasic BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, >=99.0% (titration) | Purity: >=99.0% (titration) | Mol Wt: 119.98 | 7558-80-7 | MFCD00003527 | 500G