Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phosphate monobasic dihydrate, meets analytical spec. of BP, FCC, Ph. Eur., USP, E 339, 98-100.5% (calc. to the dried substance), Solstice

Catalog No. 6001834 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
6 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-018-34 1 kg
60-018-35 5 kg
60-044-545 6 x 1 kg
60-044-544 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-018-34 Supplier Solstice Supplier No. 042691KG
Add to Cart
Add to Cart

Meets analytical specification of BP, FCC, Ph. Eur., USP, E 339, 98-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H6NaO6P
Molecular Weight (g/mol) 156.006
MDL Number MFCD00149209
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
Synonym sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
PubChem CID 23673460
IUPAC Name sodium;dihydrogen phosphate;dihydrate
SMILES O.O.OP(=O)(O)[O-].[Na+]

Specifications

Quantity 1 kg
Linear Formula NaH2PO4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 156.01g/mol
Percent Purity 98 to 100.5% (calc. to the dried substance)
Chemical Name or Material Sodium Phosphate Monobasic Dihydrate, Meets analytical specification of Ph. Eur., BP, USP, FCC
EINECSNumber 231-449-2
RTECSNumber WA1900500
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.