Learn More
Description
- SPECIFICATIONS
- Appearance (Color): White to Off-White
- Appearance (Form): Crystalline Powder
- Purity: ≥ 99.0%
- pH (5% Aqueous): 8.7 - 9.3
- Identification (FTIR): Conforms to Structure
- Loss on Drying (4h @ 105°C): 10.0 - 15.0%
- Insoluble Matter: ≤ 0.2%
- Lead: ≤ 4 ppm
- Arsenic: ≤ 3 ppm
- Phosphorous: 20.0 - 24.0%
- Chloride: ≤ 140 ppm
- Fluoride: ≤ 50 ppm
- Sulfate: ≤ 150 ppm
- DNase: None Detected
- RNase: None Detected
Specifications
Specifications
| Physical Form | Solid |
| Quantity | 10 kg |
| Percent Purity | ≥ 99.0% |
| Grade | Reagent |
| Packaging | UN Plastic Pail |
| Chemical Name or Material | Sodium phosphate monobasic, monohydrate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
