Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry

Catalog No. AC447310010
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447310010 1 kg Plastic bottle
AC447312500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447310010 Supplier Thermo Scientific Chemicals Supplier No. 447310010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 10049-21-5 | H5NaO5P | 139.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
IUPAC Name sodium;dihydrogen phosphate;hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

Melting Point 100.0°C
Quantity 1 kg
Assay Percent Range 99.5%
Linear Formula NaH2PO4·H2O
Merck Index 15,879
Solubility Information Clear colorless
Formula Weight 137.99
Percent Purity 99.5%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Sodium phosphate, monobasic monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.