Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Monobasic Monohydrate, ACS Reagent (Sodium Dihydrogen Phosphate), Reagents

Catalog No. 01337702
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-702 100 g
01-337-701 3 kg
01-337-700 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-702 Supplier Reagents Holdings Llc Supplier No. C1323500100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Phosphate Monobasic Monohydrate, ACS Reagent (Sodium Dihydrogen Phosphate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Phosphate Monobasic Monohydrate, ACS Reagent (Sodium Dihydrogen Phosphate)
Boiling Point 204°C
Color Colorless/White
CAS 10049-21-5
Physical Form Solid
Quantity 100 g
Molecular Formula NaH2PO4, H2O
Synonym Sodium Dihydrogen Phosphate Monohydrate
Molecular Weight (g/mol) 137.9923
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.04
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.