Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate, Monobasic, Monohydrate, Crystal, U.S.P. - F.C.C., J.T. Baker™

Catalog No. 02004214 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-214 500 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-004-214 Supplier Avantor Supplier No. 382001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
IUPAC Name sodium;dihydrogen phosphate;hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 137.99
Grade FCC/USP
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.04kg
Chemical Name or Material Sodium Phosphate, Monobasic, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.