Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Monobasic, Monohydrate, MP Biomedicals™

Catalog No. MP219920201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219920201 1 kg
MP21992025 500 g
MP219920205 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219920201 Supplier MP Biomedicals, Inc Supplier No. 0219920201
Add to Cart
Add to Cart

Features a very high buffering capacity. Sodium Phosphate Monobasic, Monohydrate, MP Biomedicals is a reagent with a variety of laboratory, industrial and manufacturing uses. Most neutral sodium phosphate buffer solutions consist of mixtures of Sodium Pho

Sodium phosphate occurs in several forms: monobasic (NaH2PO4), dibasic (Na2HPO4), and tribasic (Na3PO4). Most neutral sodium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.

  • A table for preparation of 0.1M sodium phosphate buffer at 25°C using various proportions of sodium phosphate monobasic and sodium phosphate dibasic has been published.
  • A study of the effect of freeze-thaw storage cycles on proteins in sodium phosphate and potassium phosphate buffer solutions has been reported.
  • The effect of 5mM sodium phosphate on the efficacy of electrospray ionization (ESI) ion mobility spectrometry (IMS) analysis has been evaluated.
  • A protocol for the purification of pyrogen-free mouse IgG1 monoclonal antibodies which uses 10 mM sodium phosphate (pH 7.4) has been published.
  • An ion-pairing HPLC method for the analysis of 5-aminosalicylic acid has been reported.
  • A TLC method for separation of nucleotide sugars in the study of glycosyltransferase activity has been published.

Sodium phosphate monobasic, monohydrate has a wide variety of uses:

  • Sequestrant, emulsifier and buffer in foods
  • Mordant in dyeing
  • Weighting silk in tanning
  • Manufacture of enamels, ceramics, detergents, boiler compounds
  • Fireproofing agent
  • In soldering and brazing instead of borax
  • Reagent and buffer in analytical chemistry
  • Cathartic
  • Laxative
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
SMILES O.[Na+].OP(O)(O)=O

Specifications

Physical Form Powder
Quantity 1 kg
Assay Percent Range 98.0–103.0%
Solubility Information Soluble in water (100mg/mL), yielding a clear, colorless solution.
IUPAC Name sodium phosphoric acid hydrate
Formula Weight 137.99g/mol
Grade USP
Recommended Storage Store at Room Temperature (15–30°C), desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.