Learn More
Alkali Scientific Sodium Phosphate Monobasic Monohydrate [Sodium dihydrogen phosphate monohydrate], 1 Kg
SDP
Catalog No. 5000660178
Encompass_Preferred
Sodium Phosphate Monobasic Monohydrate, 1 Kg is a high-quality reagent used in laboratory settings for preparing buffer solutions and adjusting pH levels. This monohydrate form of sodium phosphate monobasic is particularly useful for experiments that require precise control over pH conditions. It is commonly used in biochemical assays, molecular biology research, and pharmaceutical testing where maintaining an acidic environment is critical. The 1 Kg quantity is ideal for smaller-scale laboratory applications, providing sufficient reagent for medium-scale projects and experiments. Sodium phosphate monobasic plays a crucial role in preparing solutions for protein analysis, cell culture, and chemical synthesis.
Catalog No. 50-006-60178 Supplier Alkali Scientific Supplier No. EMO001CP
Sodium Phosphate Monobasic Monohydrate, 1 Kg is a high-quality reagent used in laboratory settings for preparing buffer solutions and adjusting pH levels. This monohydrate form of sodium phosphate monobasic is particularly useful for experiments that require precise control over pH conditions. It is commonly used in biochemical assays, molecular biology research, and pharmaceutical testing where maintaining an acidic environment is critical. The 1 Kg quantity is ideal for smaller-scale laboratory applications, providing sufficient reagent for medium-scale projects and experiments. Sodium phosphate monobasic plays a crucial role in preparing solutions for protein analysis, cell culture, and chemical synthesis.