Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Monobasic Monohydrate, Ultrapure, 500 g, Innovating Science™
GREENER_CHOICESCI_ED

Catalog No. S000973 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000973 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000973 Supplier Aldon Corporation Supplier No. IS29048
Add to Cart
Add to Cart
  • Appearance (Color): White to Off-White
  • Appearance (Form): Crystalline Powder
  • Purity: ≥ 99.0%
  • pH (5% Aqueous): 8.7 to 9.3
  • Identification (FTIR): Conforms to Structure
  • Loss on Drying (4h at 105 °C): 10.0 to 15.0%
  • Insoluble Matter: ≤ 0.2%
  • Lead: ≤ 4 ppm
  • Arsenic: ≤ 3 ppm
  • Phosphorous: 20.0 to 24.0%
  • Chloride: ≤ 140 ppm
  • Fluoride: ≤ 50 ppm
  • Sulfate: ≤ 150 ppm
  • DNase: None Detected
  • RNase: None Detected

Chemical Identifiers

CAS 10049-21-5
Molecular Weight (g/mol) 137.99

Specifications

Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Percent Purity 100%
Grade Ultra Pure
Packaging HDPE
Chemical Name or Material Sodium phosphate, monobasic, monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.