Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Solution, Saturated, for Chlorine Dioxide, LabChem™

Catalog No. LC247802 Shop All LabChem Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC247802 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC247802 Supplier LabChem, Inc. Supplier No. LC247802
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7558-79-4 , 7732-18-5
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color Colorless
CAS Max % 88, 12
Linear Formula Na2HPO4
Solubility Information Soluble in water
Formula Weight 141.96
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Phosphorus oxides
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Sodium Phosphate Solution, Saturated, for Chlorine Dioxide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes eye irritation.
Precautionary Statement GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.