Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Tribasic, 0.2 Molar, Reagents

Catalog No. 01333550
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-550 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-550 Supplier Reagents Holdings Llc Supplier No. CS1859004F
Add to Cart
Add to Cart

Sodium Phosphate Tribasic, 0.2 Molar, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 10101-89-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Trisodium Phosphate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
pH 12
Physical Form Liquid
Quantity 4 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.0287
Chemical Name or Material Sodium Phosphate Tribasic, 0.2 Molar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.