Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sodium phosphate, tribasic dodecahydrate, 98%, for biochemistry

Catalog No. AC447960025
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447960025 2.5 kg
AC447965000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447960025 Supplier Thermo Scientific Chemicals Supplier No. 447960025
Only null left
Add to Cart

CAS: 10101-89-0 | Na3O412H2O | 380.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-89-0
Molecular Formula Na3O412H2O
Molecular Weight (g/mol) 380.13
InChI Key ASTWEMOBIXQPPV-UHFFFAOYSA-K
Synonym trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
PubChem CID 61473
IUPAC Name trisodium;phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Melting Point 75.0°C
Quantity 2.5 kg
Assay Percent Range 98%
Linear Formula Na3PO4·12H2O
Merck Index 15,8792
Solubility Information Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 380.13
Percent Purity 98%
Grade Biochemical
Specific Gravity 1.62
Density 1.6200g/mL
Chemical Name or Material Sodium phosphate, tribasic dodecahydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.