Learn More
Description
Sodium Phosphate Dibasic USP is a white powder that readily absorbs moisture. It is hygroscopic.
Alternative name(s): dibasic sodium phosphate, disodium hydrogen phosphate
Specifications
Specifications
| Chemical Name or Material | Sodium Phosphate USP (Anhydrous) (Dibasic) |
| Packaging | Jar, 2.8L, White, HDPE, Round, 110-400, TE Pano Neck |
| CAS | 7558-79-4 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 141.96 |
| Quantity | 2.5 kg |
| Percent Purity | 98 to 100.5 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
