Learn More
Medchemexpress LLC Sodium phosphite dib 100g
SDP
Catalog No. 5000206352
Encompass_Preferred
Sodium phosphite dibasic pentahydrate 98% is a metal chelating agent used to remove copper ions from water supplies
Catalog No. 50-002-06352 Supplier Medchemexpress LLC Supplier No. HYW111522100G
Sodium phosphite dibasic pentahydrate 98% is a metal chelating agent used to remove copper ions from water supplies