Learn More
Medchemexpress LLC Sodium phosphite dib 250g
SDP
Catalog No. 5000206022
Encompass_Preferred
Sodium phosphite dibasic pentahydrate 98% is a metal chelating agent used to remove copper ions from water supplies
Catalog No. 50-002-06022 Supplier Medchemexpress LLC Supplier No. 250GHYW111522
Sodium phosphite dibasic pentahydrate 98% is a metal chelating agent used to remove copper ions from water supplies