Learn More
Medchemexpress LLC Sodium phosphite dib 500g
SDP
Catalog No. 5000206424
Encompass_Preferred
Sodium phosphite dibasic pentahydrate 98% is a metal chelating agent used to remove copper ions from water supplies
Catalog No. 50-002-06424 Supplier Medchemexpress LLC Supplier No. HYW111522500G
Sodium phosphite dibasic pentahydrate 98% is a metal chelating agent used to remove copper ions from water supplies