Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Phosphite Dibasic Pentahydrate

Catalog No. 04283250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-283-250G 250 g
04-283-1KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04-283-250G Supplier Sigma Aldrich Supplier No. 04283250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium phosphite dibasic pentahydrate can be used as an electron donor for the enantioselective transformation of prochiral sulfides to the corresponding optically active sulfoxides catalyzed by Thermocrispum municipale cyclohexanone monooxygenase (TmCHMO) enzyme.

Specifications

Quantity 250 g
Molecular Formula HNa2O3P -+ 5H2O
Linear Formula Na2HPO3 -+ 5H2O
MDL Number MFCD00150354
Synonym di-Sodium hydrogen phosphite
Molecular Weight (g/mol) 216.04 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.