Learn More
Selleck Chemical LLC Sodium phytate hydrate
SDP
Catalog No. 5000003358
Encompass_Preferred
Sodium phytate hydrate (Phytic acid Dodecasodium phytate Myo-inositol hexakis Inositol hexakisphosphate) functions as a storage depot and precursor for other inositol phosphates and pyrophosphates It can be used clinically as a hypocalcemic agent and a complexing agent for removal of traces of heavy metal ions
Catalog No. 50-000-03358 Supplier Selleck Chemical LLC Supplier No. S631525MG
Sodium phytate hydrate (Phytic acid Dodecasodium phytate Myo-inositol hexakis Inositol hexakisphosphate) functions as a storage depot and precursor for other inositol phosphates and pyrophosphates It can be used clinically as a hypocalcemic agent and a complexing agent for removal of traces of heavy metal ions