Learn More
Selleck Chemical LLC Sodium picosulfate monohydrate E4854-25MG
SDP
Catalog No. 5000041504
Encompass_Preferred
Sodium picosulfate monohydrate is a polyphenolic stimulant laxative and prodrug form of desacetyl bisacodyl (DAB) It is used for treating idiopathic constipation and preparing the bowel before colonoscopy
Catalog No. 50-000-41504 Supplier Selleck Chemical LLC Supplier No. E485425MG
Sodium picosulfate monohydrate is a polyphenolic stimulant laxative and prodrug form of desacetyl bisacodyl (DAB) It is used for treating idiopathic constipation and preparing the bowel before colonoscopy