Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium pivalate hydrate, 98%

Catalog No. AC364710500
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC364710500 50 g
AC364710100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC364710500 Supplier Thermo Scientific Chemicals Supplier No. 364710500
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 143174-36-1 | C5H9NaO2 | 124.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143174-36-1
Molecular Formula C5H9NaO2
Molecular Weight (g/mol) 124.12
MDL Number MFCD00150782
InChI Key SJRDNQOIQZOVQD-UHFFFAOYSA-M
Synonym sodium trimethylacetate hydrate, sodium pivalate hydrate, sodium hydrate pivalate, propanoic acid, 2,2-dimethyl-, sodium salt, hydrate, acmc-20ajn9, sodiumtrimethylacetatehydrate, c5h9o2.na.h2o, trimethylacetic acid sodium salt hydrate, sodium 2,2-dimethylpropanoate-water 1/1/1
PubChem CID 23666172
IUPAC Name sodium;2,2-dimethylpropanoate;hydrate
SMILES [Na+].CC(C)(C)C([O-])=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula (CH3)3CCO2Na·xH2O
Quantity 50 g
Formula Weight 142.13
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Sodium pivalate hydrate
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.