Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Pivalate Hydrate 98.0+%, TCI America™
SDP

Catalog No. S097825G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. S097825G

Supplier: TCI America S097825G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 143174-36-1
Molecular Formula C5H9NaO2
Molecular Weight (g/mol) 124.12
MDL Number MFCD00150782
InChI Key SJRDNQOIQZOVQD-UHFFFAOYSA-M
Synonym sodium trimethylacetate hydrate, sodium pivalate hydrate, sodium hydrate pivalate, propanoic acid, 2,2-dimethyl-, sodium salt, hydrate, acmc-20ajn9, sodiumtrimethylacetatehydrate, c5h9o2.na.h2o, trimethylacetic acid sodium salt hydrate, sodium 2,2-dimethylpropanoate-water 1/1/1
PubChem CID 23666172
IUPAC Name sodium 2,2-dimethylpropanoate
SMILES [Na+].CC(C)(C)C([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 124.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Pivalate Hydrate
RTECSNumber UA2459665
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.