Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Polyacrylate
SCI_ED

Catalog No. S25565 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Solid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25565 100 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25565

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is an Educational Grade Chemical -- no C of A is available.

Chemical Identifiers

CAS 9003-04-7 , 9003-01-4
Molecular Formula C3H3NaO2
Molecular Weight (g/mol) 94.05
InChI Key NNMHYFLPFNGQFZ-UHFFFAOYSA-M
Synonym sodium acrylate, sodium prop-2-enoate, sodium polyacrylate, 2-propenoic acid, sodium salt, polyco, rhotex gs, unii-7c98fkb43h, acrysol lmw-45n, hiviswako 105, sodium poly acrylate
PubChem CID 4068533
IUPAC Name sodium;prop-2-enoate
SMILES C=CC(=O)[O-].[Na+]

Specifications

Linear Formula (C3H3NaO2)n
Density 0.4 to 0.7g/mL
Quantity 100 g
Packaging Solid
Chemical Name or Material Sodium Polyacrylate
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.