Learn More
Educational Innovations Sodium Polyacrylate, MFCD00147949, 2270g (5lb package)
SDP
Supplier:  Educational Innovations GB620
Encompass
This fine white powder instantly turns liquid water into a slush-like solid substance. Technically speaking, the polymer absorbs from 500 to 1,000 times its own weight in water. 2270g (5lb package)This is the super-absorbent polymer found in disposable baby diapers. Also used in many "disappearing water" magic tricks, this fine white powder instantly turns liquid water into a slush-like solid substance. Technically speaking, the polymer absorbs from 500 to 1,000 times its own weight in water. Simply add table salt to reverse the reaction.
Catalog No. NC0045734