Learn More
Medchemexpress LLC Sodium polyphosphate 500g
SDP
Catalog No. 5000206126
Encompass_Preferred
Sodium polyphosphate crystal 200 mesh 96% is a water-soluble inorganic stabilizer with surfactant and metal ion chelator activity Sodium polyphosphate crystal 200 mesh 96% can be widely used in food cosmetics and industrial products to improve product texture and stability Sodium polyphosphate crystal 200 mesh 96% also helps prevent metal ion-induced degradation and extend the product s life
Catalog No. 50-002-06126 Supplier Medchemexpress LLC Supplier No. HYW111692500G
Sodium polyphosphate crystal 200 mesh 96% is a water-soluble inorganic stabilizer with surfactant and metal ion chelator activity Sodium polyphosphate crystal 200 mesh 96% can be widely used in food cosmetics and industrial products to improve product texture and stability Sodium polyphosphate crystal 200 mesh 96% also helps prevent metal ion-induced degradation and extend the product s life