Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Polyphosphates, Glassy, FCC, 60-71%, Spectrum™ Chemical
SDP

Catalog No. 18606744 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-744 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-606-744 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO169500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SO169, 68915-31-1, Approx. (NaPO3)n

Spectrum™ Chemical Sodium Polyphosphates, Glassy, FCC is used as an acidity regulator, emulsifier and buffering agent, and as a sequestering agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 68915-31-1
Molecular Formula H2Na2O7P2
Molecular Weight (g/mol) 221.94
InChI Key GYQBBRRVRKFJRG-UHFFFAOYSA-L
IUPAC Name disodium hydrogen (hydrogen phosphonatooxy)phosphonate
SMILES [Na+].[Na+].OP([O-])(=O)OP(O)([O-])=O

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 500 g
Percent Purity 60 to 71%
Grade FCC
Packaging Poly Bottle
Chemical Name or Material Sodium Polyphosphates, Glassy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.