Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Polytungstate Solution, ∼85% in H2O, For separation of minerals, MilliporeSigma™ Supelco™

Catalog No. 111012770 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2770 100 mL
11-101-2771 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2770 Supplier MilliporeSigma Supelco Supplier No. 80656100ML
Add to Cart
Add to Cart

Specifications

CAS 12141-67-2
Quantity 100 mL
Linear Formula Na6O39W12
MDL Number MFCD00061466
Formula Weight 2968.00
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.